Gene Transcripts

Gene Symbol
DKK3
Entrez Gene ID
27122
Gene Name
dickkopf WNT signaling pathway inhibitor 3
Gene Aliases
REIC, RIG
Chromosome Location
Chr. 11: 11962996 - 12009370 on Build GRCh38
UniGene ID
Hs.292156
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018057.1 NP_001018067.1 2, 3 405
NM_013253.4 NP_037385.2 3, 4 573
NM_015881.5 NP_056965.3 3, 4 587
XM_006718178.2 XP_006718241.1 3, 4 400
XM_017017553.1 XP_016873042.1 3, 4 534
XM_017017554.1 XP_016873043.1 3, 4 552
XM_017017555.1 XP_016873044.1 3, 4 400
GenBank mRNA AB033421.1 - 2, 3 366
AB034203.1 - 3, 4 545
AB057591.1 - 3, 4 573
AF177396.1 - 2, 3 385
AK090952.1 - 492
AK092979.1 - 3, 4 364
AK225156.1 - 2, 3 357
AK289897.1 - 3, 4 414
AK295539.1 - 3, 4 508
AL535720.3 - 2, 3 404
AY358378.1 - 3, 4 500
AY587550.1 - 2, 3 348
BC007660.2 - 3, 4 586 3347453

View Full Product Details