Gene Transcripts

Gene Symbol
IQCE
Entrez Gene ID
23288
Gene Name
IQ motif containing E
Gene Aliases
1700028P05Rik
Chromosome Location
Chr. 7: 2558972 - 2614734 on Build GRCh38
UniGene ID
Hs.520627
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287499.1 NP_001274428.1 6 648
NM_001287500.1 NP_001274429.1 5 600
NM_001287501.1 NP_001274430.1 4 554
NM_001287502.1 NP_001274431.1 4 554
NM_152558.4 NP_689771.3 6 648
XM_006715676.2 XP_006715739.1 4 402
XM_011515242.1 XP_011513544.1 5 507
XM_011515243.1 XP_011513545.1 4 529
XM_017011902.1 XP_016867391.1 6 710
XM_017011903.1 XP_016867392.1 6 588
GenBank mRNA AB028946.2 - 4 351
AK092404.1 - 6 542
AK293372.1 - 5 423
AK302053.1 - 4 502
AK309646.1 - 6 572
AL136792.1 - 5 329
BC043150.1 - 4 474 5297871
BC061518.1 - 5 484
BC071858.1 - 6 622 5478116
KJ534872.1 - 6 438
KJ535081.1 - 4 344

View Full Product Details