Gene Transcripts

Gene Symbol
SYVN1
Entrez Gene ID
84447
Gene Name
synoviolin 1
Gene Aliases
DER3, HRD1
Chromosome Location
Chr. 11: 65127279 - 65135178 on Build GRCh38
UniGene ID
Hs.75859
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032431.2 NP_115807.1 3 249
NM_172230.2 NP_757385.1 3 249
XM_011545303.2 XP_011543605.1 3 1097
GenBank mRNA AB058713.1 - 3 197
AB085847.1 - 2 155
AF317634.1 - 2 155
AK074148.1 - 3 204
AK126912.1 - 3 250
AL834262.1 - 3 249
BC030530.1 - 3 190 5211742

View Full Product Details