Gene Transcripts

Gene Symbol
HIGD1B
Entrez Gene ID
51751
Gene Name
HIG1 hypoxia inducible domain family member 1B
Gene Aliases
CLST11240, CLST11240-15
Chromosome Location
Chr. 17: 44844281 - 44850480 on Build GRCh38
UniGene ID
Hs.151787
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271880.1 NP_001258809.1 2 247
NM_016438.3 NP_057522.1 1 331
NR_073504.1 1 331
XM_006721946.2 XP_006722009.1 2 225
XM_006721947.3 XP_006722010.1 2 635
XM_006721948.3 XP_006722011.1 2 329
XM_011524891.2 XP_011523193.1 2 729
XM_017024742.1 XP_016880231.1 2 675
XM_017024743.1 XP_016880232.1 2 767
GenBank mRNA AB038021.1 - 1 158
AB038022.1 - 1 158
BC020667.1 - 1 322 4724613
BG573492.1 - 1 323
BM503772.1 - 1 334
BM876435.1 - 1 307

View Full Product Details