Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016602.2 | NP_057686.2 | 2 | 1082 | |
GenBank mRNA | AF208237.1 | - | 2 | 1075 | |
AF215981.1 | - | 2 | 1082 | ||
AY429103.1 | - | 2 | 1078 | ||
BC098132.1 | - | 2 | 1082 | 40004874 | |
BC098157.1 | - | 2 | 1082 | 40004879 | |
BC098304.1 | - | 2 | 1082 | 40004880 | |
BC099840.1 | - | 2 | 1082 | 40004873 | |
BM930170.1 | - | 1 | 436 |