Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136201.1 | NP_001129673.1 | 3 | 428 | |
NM_024710.2 | NP_078986.1 | 3 | 428 | ||
GenBank mRNA | AK027122.1 | - | 3 | 391 | |
AK311540.1 | - | 1068 | |||
BC017344.2 | - | 3 | 315 | 4893552 | |
BP226043.1 | - | 3 | 429 | ||
CR457377.1 | - | 2 | 254 |