Gene Transcripts

Gene Symbol
COMTD1
Entrez Gene ID
118881
Gene Name
catechol-O-methyltransferase domain containing 1
Gene Aliases
Chromosome Location
Chr. 10: 75233971 - 75236012 on Build GRCh38
UniGene ID
Hs.355333
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144589.2 NP_653190.2 7 782
GenBank mRNA AK074421.1 - 7 735
AK290452.1 - 7 836
AK302283.1 - 7 692
AY358476.1 - 7 781
BC023663.2 - 7 703 4897624
BC047774.1 - 7 782 5761696

View Full Product Details