Gene Transcripts

Gene Symbol
COX7A1
Entrez Gene ID
1346
Gene Name
cytochrome c oxidase subunit 7A1
Gene Aliases
COX7A, COX7AH, COX7AM
Chromosome Location
Chr. 19: 36150922 - 36152869 on Build GRCh38
UniGene ID
Hs.421621
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001864.3 NP_001855.1 2 554
GenBank mRNA AK312091.1 - 2 393
BC002757.2 - 2 554 3631740
BT006924.1 - 2 92
CR542120.1 - 2 92
M83186.1 - 2 133

View Full Product Details