Gene Transcripts

Gene Symbol
Ins2
Entrez Gene ID
24506
Gene Name
insulin 2
Gene Aliases
Chromosome Location
Chr. 1: 215856967 - 215858034 on Build Rnor_6.0
UniGene ID
Rn.989
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019130.2 NP_062003.1 2 200
GenBank mRNA BP485158.1 - 2 199

View Full Product Details