Gene Transcripts

Gene Symbol
Kiss1r
Entrez Gene ID
78976
Gene Name
KISS1 receptor
Gene Aliases
Gpr54
Chromosome Location
Chr. 7: 12606347 - 12609694 on Build Rnor_6.0
UniGene ID
Rn.48681
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301151.1 NP_001288080.1 2 444
NM_023992.2 NP_076482.2 2 444
GenBank mRNA AB051066.1 - 2 270
AF115516.1 - 2 270

View Full Product Details