Gene Transcripts

Gene Symbol
Rln3
Entrez Gene ID
266997
Gene Name
relaxin 3
Gene Aliases
Insl7
Chromosome Location
Chr. 19: 25164497 - 25166528 on Build Rnor_6.0
UniGene ID
Rn.45123
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170667.2 NP_733767.1 2 236
GenBank mRNA AB076564.1 - 2 236
AY112741.1 - 2 236

View Full Product Details