Gene Transcripts
- Gene Symbol
- Prl8a5
- Entrez Gene ID
- 286889
- Gene Name
- prolactin family 8, subfamily a, member 5
- Gene Aliases
- Chromosome Location
- Chr. 17: 39411716 - 39417929 on Build Rnor_6.0
- UniGene ID
- Rn.3757
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173110.1 |
NP_775133.1
|
2, 3 |
240 |
|
GenBank mRNA |
U09099.1 |
- |
2, 3 |
240 |
|
View Full Product Details