Gene Transcripts

Gene Symbol
CUL1
Entrez Gene ID
8454
Gene Name
cullin 1
Gene Aliases
Chromosome Location
Chr. 7: 148698178 - 148801110 on Build GRCh38
UniGene ID
Hs.146806
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003592.2 NP_003583.2 3, 4 673
XM_005250060.4 XP_005250117.1 3, 4 709
XM_011516629.2 XP_011514931.1 3, 4 665
XM_011516630.2 XP_011514932.1 3, 4 566
XM_011516631.2 XP_011514933.1 3, 4 694
XM_011516632.2 XP_011514934.1 3, 4 592
XM_017012723.1 XP_016868212.1 3, 4 593
GenBank mRNA AF062536.1 - 2, 3 447
AK096163.1 - 4, 5 561
AK315023.1 - 3, 4 849
BC125119.1 - 3, 4 586 40118923
BC125120.1 - 3, 4 586 40118927
BX537409.1 - 3, 4 603
U58087.1 - 3, 4 375

View Full Product Details