Gene Transcripts

Gene Symbol
C4bpa
Entrez Gene ID
24235
Gene Name
complement component 4 binding protein, alpha
Gene Aliases
C4BP
Chromosome Location
Chr. 13: 47342090 - 47377580 on Build Rnor_6.0
UniGene ID
Rn.10408
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012516.2 NP_036648.2 5 634
GenBank mRNA BC088165.1 - 5 634
Z50051.1 - 3 297

View Full Product Details