Gene Transcripts

Gene Symbol
Slc2a2
Entrez Gene ID
25351
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 2
Gene Aliases
GTT2, Glut2
Chromosome Location
Chr. 2: 114413431 - 114445418 on Build Rnor_6.0
UniGene ID
Rn.89295
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012879.2 NP_037011.2 3, 4 406
XM_006232207.2 XP_006232269.1 4, 5 710
GenBank mRNA BC078875.1 - 3, 4 406
J03145.1 - 3, 4 481

View Full Product Details