Gene Transcripts

Gene Symbol
Cacna1a
Entrez Gene ID
25398
Gene Name
calcium channel, voltage-dependent, P/Q type, alpha 1A subunit
Gene Aliases
BccA1, Cav2.1, rbA-1
Chromosome Location
Chr. 19: 25453195 - 25749550 on Build Rnor_6.0
UniGene ID
Rn.87769
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012918.3 NP_037050.2 37 5411
XM_008772347.1 XP_008770569.1 37 5862
XM_008772349.1 XP_008770571.1 37 5859
XM_008772350.1 XP_008770572.1 36 5856
XM_008772351.1 XP_008770573.1 36 5850
XM_008772352.1 XP_008770574.1 37 5862
XM_008772353.1 XP_008770575.1 38 5754
XM_008772355.1 XP_008770577.1 37 5748
XM_008772357.1 XP_008770579.1 36 5736
XM_008772360.1 XP_008770582.1 37 5862
GenBank mRNA M64373.1 - 37 5411

View Full Product Details