Gene Transcripts

Gene Symbol
Esp23
Entrez Gene ID
100126779
Gene Name
exocrine gland secreted peptide 23
Gene Aliases
Gm20581
Chromosome Location
Chr. 17: 39073691 - 39077037 on Build GRCm38
UniGene ID
Mm.471589
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177582.1 NP_001171053.1 3 342
GenBank mRNA AB307002.1 - 3 342

View Full Product Details