Gene Transcripts

Gene Symbol
Vmn1r9
Entrez Gene ID
171203
Gene Name
vomeronasal 1 receptor 9
Gene Aliases
V1rc30
Chromosome Location
Chr. 6: 57070895 - 57071945 on Build GRCm38
UniGene ID
Mm.377181
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134185.2 NP_598946.2 1 487
GenBank mRNA BC132241.1 - 1 487
BC132577.1 - 1 487 40130854

View Full Product Details