Gene Transcripts

Gene Symbol
Ntrk1
Entrez Gene ID
18211
Gene Name
neurotrophic tyrosine kinase, receptor, type 1
Gene Aliases
C80751, Tkr, TrkA, trk
Chromosome Location
Chr. 3: 87778244 - 87795242 on Build GRCm38
UniGene ID
Mm.80682
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033124.1 NP_001028296.1 14 1786
GenBank mRNA AK081588.1 - 14 1789
AK148691.1 - 7 695
BC051596.1 - 4 480

View Full Product Details