Gene Transcripts
- Gene Symbol
- Prl3d2
- Entrez Gene ID
- 215028
- Gene Name
- prolactin family 3, subfamily d, member 1
- Gene Aliases
- PL-Ib, Plib
- Chromosome Location
- Chr. 13: 27121633 - 27127555 on Build GRCm38
- UniGene ID
- Mm.458451
- Species
- Mus musculus
- Gene Symbol
- Prl3d1
- Entrez Gene ID
- 18775
- Gene Name
- prolactin family 3, subfamily d, member 1
- Gene Aliases
- AI325057, Csh1, PL-I, PL-I-alpha, PL-Ia, Pl-1, Pl1, mPL-I
- Chromosome Location
- Chr. 13: 27094189 - 27100260 on Build GRCm38
- UniGene ID
- Mm.233151
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF525162.1 |
- |
|
122 |
|
View Full Product Details