Gene Transcripts

Gene Symbol
Cdh19
Entrez Gene ID
227485
Gene Name
cadherin 19, type 2
Gene Aliases
Chromosome Location
Chr. 1: 110888002 - 110977593 on Build GRCm38
UniGene ID
Mm.322070
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081386.1 NP_001074855.1 11 1850
GenBank mRNA BC157927.1 - 11 1841

View Full Product Details