Gene Transcripts

Gene Symbol
Cdr2l
Entrez Gene ID
237988
Gene Name
cerebellar degeneration-related protein 2-like
Gene Aliases
D030068L24Rik, Gm21
Chromosome Location
Chr. 11: 115381916 - 115396132 on Build GRCm38
UniGene ID
Mm.102872
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080929.1 NP_001074398.1 5 3614
GenBank mRNA AK051090.1 - 1 1413

View Full Product Details