Gene Transcripts

Gene Symbol
Ntn5
Entrez Gene ID
243967
Gene Name
netrin 5
Gene Aliases
Gm484
Chromosome Location
Chr. 7: 45684022 - 45694556 on Build GRCm38
UniGene ID
Mm.291240
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033356.3 NP_001028528.2 2 684
NM_001289692.1 NP_001276621.1 2 684
GenBank mRNA AK142898.1 - 2 292
BC150901.1 - 2 277 9087812
BC172055.1 - 2 277

View Full Product Details