Gene Transcripts

Gene Symbol
Trank1
Entrez Gene ID
320429
Gene Name
tetratricopeptide repeat and ankyrin repeat containing 1
Gene Aliases
A230061D21Rik, C030048J01Rik, Gm187, Lba1
Chromosome Location
Chr. 9: 111311590 - 111395775 on Build GRCm38
UniGene ID
Mm.270754
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164659.1 NP_001158131.1 20 6661
GenBank mRNA AK129119.1 - 8 2726
BC086653.1 - 8 1687 30536238

View Full Product Details