Gene Transcripts

Gene Symbol
Cyb5d1
Entrez Gene ID
327951
Gene Name
cytochrome b5 domain containing 1
Gene Aliases
Gm740
Chromosome Location
Chr. 11: 69390930 - 69395346 on Build GRCm38
UniGene ID
Mm.415669
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045525.1 NP_001038990.1 4 670
GenBank mRNA BC099556.1 - 4 670

View Full Product Details