Gene Transcripts

Gene Symbol
Dok6
Entrez Gene ID
623279
Gene Name
docking protein 6
Gene Aliases
Dok-6
Chromosome Location
Chr. 18: 89301135 - 89769136 on Build GRCm38
UniGene ID
Mm.447164
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039173.1 NP_001034262.1 1 19
GenBank mRNA AB221044.1 - 1 19
BC138955.1 - 1 70 8861977
BC138957.1 - 1 70 8861979

View Full Product Details