Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163513.1 | NP_001156985.1 | 11 | 2436 | |
NM_027726.1 | NP_082002.1 | 10 | 2367 | ||
GenBank mRNA | AK147598.1 | - | 10 | 1936 | |
AK147699.1 | - | 11 | 2437 | ||
BC043128.1 | - | 1 | 92 | ||
BC060142.1 | - | 3 | 317 | ||
BC067046.1 | - | 6 | 904 |