Gene Transcripts

Gene Symbol
Prr16
Entrez Gene ID
71373
Gene Name
proline rich 16
Gene Aliases
5430406M13Rik, AI607429
Chromosome Location
Chr. 18: 51117898 - 51305129 on Build GRCm38
UniGene ID
Mm.63546
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081224.2 NP_001074693.1 2 526
GenBank mRNA AK017282.1 - 1 340
AK042678.1 - 687
AK045646.1 - 1 480
AK046456.1 - 2 691
AK081911.1 - 502
BC132481.1 - 2 526 40130758

View Full Product Details