Gene Transcripts

Gene Symbol
Ddrgk1
Entrez Gene ID
77006
Gene Name
DDRGK domain containing 1
Gene Aliases
1110001I20Rik, 2600009E05Rik, AI326138, Ufbp1
Chromosome Location
Chr. 2: 130654083 - 130665600 on Build GRCm38
UniGene ID
Mm.440063
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029832.2 NP_084108.1 5 651
GenBank mRNA BC051541.1 - 3 240
BC115616.2 - 5 651 40102754
BC115617.1 - 5 651 40102757

View Full Product Details