Gene Transcripts

Gene Symbol
Qser1
Entrez Gene ID
99003
Gene Name
glutamine and serine rich 1
Gene Aliases
4732486I23Rik, AI502974
Chromosome Location
Chr. 2: 104754793 - 104816841 on Build GRCm38
UniGene ID
Mm.274314
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001123327.2 NP_001116799.1 8 4776
GenBank mRNA AK141733.1 - 6 3087
CF749097.1 - 5 686

View Full Product Details