Gene Transcripts

Gene Symbol
Lrrn3
Entrez Gene ID
81514
Gene Name
leucine rich repeat neuronal 3
Gene Aliases
Nlrr3
Chromosome Location
Chr. 6: 61374328 - 61405195 on Build Rnor_6.0
UniGene ID
Rn.39124
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030856.1 NP_110483.1 2 1312
GenBank mRNA AF291437.1 - 2 1312
BC081791.1 - 1 658 7124169

View Full Product Details