Gene Transcripts
- Gene Symbol
- BNIP1
- Entrez Gene ID
- 662
- Gene Name
- BCL2 interacting protein 1
- Gene Aliases
- NIP1, SEC20, TRG-8
- Chromosome Location
- Chr. 5: 173144442 - 173164387 on Build GRCh38
- UniGene ID
- Hs.145726
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001205.2 |
NP_001196.2
|
1, 2 |
196 |
|
NM_013978.2 |
NP_053581.2
|
1, 2 |
196 |
|
NM_013979.2 |
NP_053582.2
|
1, 2 |
196 |
|
NM_013980.2 |
NP_053583.2
|
1, 2 |
196 |
|
XM_011534638.1 |
XP_011532940.1
|
1, 2 |
191 |
|
XM_011534639.1 |
XP_011532941.1
|
1, 2 |
191 |
|
GenBank mRNA |
AF083956.1 |
- |
1, 2 |
92 |
|
AF083957.1 |
- |
1, 2 |
102 |
|
AF083958.1 |
- |
1, 2 |
102 |
|
AU311051.1 |
- |
1, 2 |
196 |
|
AY216799.1 |
- |
1, 2 |
92 |
|
BC010959.1 |
- |
1, 2 |
121 |
4052817 |
U15172.1 |
- |
1, 2 |
102 |
|
View Full Product Details