Gene Transcripts

Gene Symbol
NR5A1
Entrez Gene ID
2516
Gene Name
nuclear receptor subfamily 5 group A member 1
Gene Aliases
AD4BP, ELP, FTZ1, FTZF1, POF7, SF-1, SF1, SPGF8, SRXY3, hSF-1
Chromosome Location
Chr. 9: 124481234 - 124507420 on Build GRCh38
UniGene ID
Hs.495108
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004959.4 NP_004950.2 7 2087
XM_005251871.3 XP_005251928.1 7 2115
XM_011518455.2 XP_011516757.1 7 1965
GenBank mRNA AK090858.1 - 2 1533
BC032501.1 - 7 2086 5227690

View Full Product Details