Gene Transcripts

Gene Symbol
Cdh17
Entrez Gene ID
117048
Gene Name
cadherin 17
Gene Aliases
Chromosome Location
Chr. 5: 25354187 - 25443675 on Build Rnor_6.0
UniGene ID
Rn.76726
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053977.2 NP_446429.1 4 300
XM_008763521.1 XP_008761743.1 4 1754

View Full Product Details