Gene Transcripts

Gene Symbol
Slc5a6
Entrez Gene ID
170551
Gene Name
solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6
Gene Aliases
SMVT
Chromosome Location
Chr. 6: 26685823 - 26697110 on Build Rnor_6.0
UniGene ID
Rn.11105
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130746.1 NP_570102.1 8 1008
GenBank mRNA AF026554.1 - 8 1008
AF081204.2 - 5 596
AF081205.1 - 5 596
BC168145.1 - 6 870 8371761

View Full Product Details