Gene Transcripts

Gene Symbol
Casp12
Entrez Gene ID
156117
Gene Name
caspase 12
Gene Aliases
Chromosome Location
Chr. 8: 2658801 - 2689184 on Build Rnor_6.0
UniGene ID
Rn.81078
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130422.1 NP_569106.1 1 246
XM_008765891.1 XP_008764113.1 2 327
GenBank mRNA AF317633.1 - 1 246
FJ465154.1 - 1 231

View Full Product Details