Gene Transcripts

Gene Symbol
Ptprh
Entrez Gene ID
171125
Gene Name
protein tyrosine phosphatase, receptor type, H
Gene Aliases
Bem2
Chromosome Location
Chr. 1: 72809937 - 72859161 on Build Rnor_6.0
UniGene ID
Rn.10285
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191945.1 NP_001178874.1 11 1919
XM_008758887.1 XP_008757109.1 14 3472
GenBank mRNA D45413.1 - 3 468

View Full Product Details