Gene Transcripts
- Gene Symbol
- Adam6
- Entrez Gene ID
- 192271
- Gene Name
- a disintegrin and metallopeptidase domain 6
- Gene Aliases
- TMDCIV
- Chromosome Location
- Chr. 6: 138432550 - 138435038 on Build Rnor_6.0
- UniGene ID
- Rn.24189
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138906.1 |
NP_620261.1
|
1 |
2231 |
|
GenBank mRNA |
Y09111.1 |
- |
1 |
2231 |
|
View Full Product Details