Gene Transcripts

Gene Symbol
Cyp4x1
Entrez Gene ID
246767
Gene Name
cytochrome P450, family 4, subfamily x, polypeptide 1
Gene Aliases
Chromosome Location
Chr. 5: 133929532 - 133959447 on Build Rnor_6.0
UniGene ID
Rn.85477
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145675.1 NP_663708.1 7 818
GenBank mRNA AF439343.1 - 7 818

View Full Product Details