Gene Transcripts
- Gene Symbol
- Cdk11b
- Entrez Gene ID
- 252879
- Gene Name
- cyclin-dependent kinase 11B
- Gene Aliases
- Cdc2l1, Cdk11
- Chromosome Location
- Chr. 5: 173051898 - 173078049 on Build Rnor_6.0
- UniGene ID
- Rn.10204
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
L24388.1 |
- |
|
635 |
|
View Full Product Details