Gene Transcripts

Gene Symbol
Eaf2
Entrez Gene ID
266787
Gene Name
ELL associated factor 2
Gene Aliases
Traits
Chromosome Location
Chr. 11: 66878007 - 66923092 on Build Rnor_6.0
UniGene ID
Rn.20681
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172047.1 NP_742044.1 2 281
XM_008768707.1 XP_008766929.1 2 168
GenBank mRNA AY049022.1 - 2 281

View Full Product Details