Gene Transcripts
- Gene Symbol
- Igfals
- Entrez Gene ID
- 79438
- Gene Name
- insulin-like growth factor binding protein, acid labile subunit
- Gene Aliases
- Als
- Chromosome Location
- Chr. 10: 14240308 - 14243554 on Build Rnor_6.0
- UniGene ID
- Rn.7327
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053329.2 |
NP_445781.2
|
2 |
251 |
|
GenBank mRNA |
S46785.1 |
- |
2 |
251 |
|
View Full Product Details