Gene Transcripts

Gene Symbol
Uts2r
Entrez Gene ID
57305
Gene Name
urotensin 2 receptor
Gene Aliases
Gpr14, Senr, UR-2-R
Chromosome Location
Chr. 10: 110364291 - 110365451 on Build Rnor_6.0
UniGene ID
Rn.44230
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020537.1 NP_065412.1 1 136
GenBank mRNA AB012210.1 - 1 136
AB029611.1 - 1 136

View Full Product Details