Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007664.4 | NP_031690.3 | 2 | 735 | |
GenBank mRNA | AB008811.1 | - | 2 | 412 | |
AK087418.1 | - | 2 | 149 | ||
AK146981.1 | - | 2 | 932 | ||
BC022107.1 | - | 2 | 166 | 5100313 | |
M31131.1 | - | 2 | 412 |