Gene Transcripts

Gene Symbol
Cbx5
Entrez Gene ID
12419
Gene Name
chromobox 5
Gene Aliases
2610029O15Rik, C75991, HP1, Hp1a, Hp1alpha
Chromosome Location
Chr. 15: 103191546 - 103239904 on Build GRCm38
UniGene ID
Mm.262059
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001076789.1 NP_001070257.1 2, 3 276
NM_001110216.1 NP_001103686.1 3, 4 377
NM_007626.3 NP_031652.1 2, 3 277
GenBank mRNA AK008792.1 - 2, 3 312
AK018349.1 - 2, 3 223
AK030366.1 - 2, 3 277
AK030442.1 - 3, 4 330
AK032975.1 - 2, 3 233
BC004707.1 - 2, 3 276 3499489
X99641.1 - 2, 3 209

View Full Product Details