Gene Transcripts

Gene Symbol
Ccne1
Entrez Gene ID
12447
Gene Name
cyclin E1
Gene Aliases
AW538188, CycE1
Chromosome Location
Chr. 7: 38097984 - 38107490 on Build GRCm38
UniGene ID
Mm.16110
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007633.2 NP_031659.2 5 443
GenBank mRNA AK089950.1 - 5 375
AK165658.1 - 5 433
BC062152.1 - 5 470 6510163
BC084588.1 - 5 487 6506897
BC106191.1 - 5 443 6336006
BC138660.1 - 4 276 8861680
BC138662.1 - 4 276 8861683
X75888.1 - 4 283

View Full Product Details