Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021138.3 | NP_066961.2 | 11 | 1502 | |
XM_011518973.2 | XP_011517275.1 | 12 | 1806 | ||
XM_011518974.2 | XP_011517276.1 | 11 | 2407 | ||
XM_011518975.2 | XP_011517277.1 | 12 | 1734 | ||
XM_011518976.2 | XP_011517278.1 | 12 | 1775 | ||
XM_011518977.2 | XP_011517279.1 | 11 | 1495 | ||
XM_011518978.2 | XP_011517280.1 | 11 | 2916 | ||
XM_017015094.1 | XP_016870583.1 | 11 | 1550 | ||
XM_017015095.1 | XP_016870584.1 | 11 | 2260 | ||
GenBank mRNA | AK054686.1 | - | 10 | 1327 | |
AK289722.1 | - | 12 | 1618 | ||
AK294607.1 | - | 11 | 1361 | ||
AK298370.1 | - | 11 | 1466 | ||
BC032410.1 | - | 10 | 1488 | 5216790 | |
BC033810.1 | - | 11 | 1490 | 5182942 | |
BC043492.1 | - | 11 | 1502 | 5767794 | |
BC064662.1 | - | 10 | 1497 | 6528158 | |
BX538160.1 | - | 10 | 1444 |