Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166368.1 | NP_001159840.1 | 6 | 464 | |
NM_008020.3 | NP_032046.1 | 6 | 479 | ||
GenBank mRNA | AK144284.1 | - | 6 | 480 | |
AK165394.1 | - | 6 | 444 | ||
BC031824.1 | - | 6 | 433 | 4196363 | |
BC053692.1 | - | 6 | 469 | 6395841 | |
CF949699.1 | - | 5 | 283 | ||
DV655383.1 | - | 6 | 464 |