Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163131.1 | NP_001156603.1 | 9 | 1790 | |
NM_007921.3 | NP_031947.1 | 9 | 1730 | ||
GenBank mRNA | AF016294.1 | - | 9 | 1696 | |
AK143135.1 | - | 9 | 1780 | ||
BC138692.1 | - | 9 | 1694 | 8861714 | |
BC145380.1 | - | 9 | 1754 | 9053938 |