Gene Transcripts

Gene Symbol
Fpr-rs3
Entrez Gene ID
14290
Gene Name
formyl peptide receptor, related sequence 3
Gene Aliases
Chromosome Location
Chr. 17: 20623846 - 20624877 on Build GRCm38
UniGene ID
Mm.455334
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008040.2 NP_032066.2 1 572
GenBank mRNA BC134352.1 - 1 567

View Full Product Details